Search
Search
| Gene Symbol: KLF15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022406293.1 | XP_022262001.1 | 993 | 104 | ||
| XM_022406294.1 | XP_022262002.1 | 944 | 104 | |||
| XM_846268.4 | XP_851361.1 | 1321 | 104 | |||
Set Membership: |
|