Search
Search
| Gene Symbol: UNC45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434376.4 | XP_003434424.1 | 4 - 5 | 503 | 125 | |
| XM_022416942.1 | XP_022272650.1 | 4 - 5 | 543 | 125 | ||
| XM_844928.5 | XP_850021.1 | 4 - 5 | 493 | 125 | ||
Set Membership: |
|