Search
Search

SLC12A6| Gene Symbol: SLC12A6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638183.3 | XP_005638240.1 | 5 - 6 | 1085 | 174 | |
| XM_005638184.3 | XP_005638241.1 | 3 - 4 | 1427 | 174 | ||
| XM_014109414.2 | XP_013964889.1 | 4 - 5 | 548 | 174 | ||
| XM_851514.5 | XP_856607.1 | 4 - 5 | 529 | 174 | ||
Set Membership: |
|