Search
Search
| Gene Symbol: MED23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003431721.4 | XP_003431769.2 | 19 - 20 | 2337 | 148 | |
| XM_022425797.1 | XP_022281505.1 | 19 - 20 | 2376 | 148 | ||
| XM_022425798.1 | XP_022281506.1 | 19 - 20 | 2364 | 148 | ||
| XM_022425799.1 | XP_022281507.1 | 18 - 19 | 2358 | 148 | ||
| XM_022425800.1 | XP_022281508.1 | 19 - 20 | 2376 | 148 | ||
| XM_022425801.1 | XP_022281509.1 | 19 - 20 | 2376 | 148 | ||
| XM_022425802.1 | XP_022281510.1 | 19 - 20 | 2376 | 148 | ||
| XM_532052.6 | XP_532052.4 | 18 - 19 | 2319 | 148 | ||
Set Membership: |
|