Search
Search

TMEM164| Gene Symbol: TMEM164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641668.3 | XP_005641725.1 | 6 - 7 | 1128 | 64 | |
| XM_005641672.2 | XP_005641729.1 | 6 - 7 | 491 | 64 | ||
| XM_005641674.3 | XP_005641731.1 | 5 - 6 | 408 | 64 | ||
| XM_022416388.1 | XP_022272096.1 | 6 - 7 | 1105 | 64 | ||
| XM_858098.5 | XP_863191.1 | 6 - 7 | 1092 | 64 | ||
Set Membership: |
|