Search
Search
| Gene Symbol: COQ6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116050.2 | XP_013971525.1 | 7 - 8 | 783 | 124 | |
| XM_022422207.1 | XP_022277915.1 | 8 - 9 | 754 | 124 | ||
| XM_547900.5 | XP_547900.1 | 8 - 9 | 918 | 124 | ||
Set Membership: |
|