Search
Search
| Gene Symbol: EOGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009187.1 | NP_001009187.1 | 1434 | 159 | ||
| XM_005632037.3 | XP_005632094.1 | 1822 | 159 | |||
| XM_005632038.3 | XP_005632095.1 | 1740 | 159 | |||
| XM_005632039.3 | XP_005632096.1 | 1834 | 159 | |||
| XM_022406254.1 | XP_022261962.1 | 1830 | 159 | |||
| XM_022406255.1 | XP_022261963.1 | 1818 | 159 | |||
| GenBank mRNA | AJ868227.1 | - | 1434 | 159 | ||
Set Membership: |
|