Search
Search
| Gene Symbol: VCAN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003434417.4 | XP_003434465.1 | 9903 | 104 | ||
| XM_022416747.1 | XP_022272455.1 | 6742 | 104 | |||
| XM_022416748.1 | XP_022272456.1 | 4525 | 104 | |||
Set Membership: |
|