Search
Search
| Gene Symbol: ABCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003081.1 | NP_001003081.1 | 27 - 28 | 3845 | 143 | |
| GenBank mRNA | AF403242.1 | - | 3 - 4 | 265 | 143 | |
| AJ315401.1 | - | 27 - 28 | 3845 | 143 | ||
| AY582532.1 | - | 27 - 28 | 3843 | 143 | ||
| Y18220.1 | - | 3845 | 143 | |||
Set Membership: |
|