Search
Search
| Gene Symbol: VDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636918.2 | XP_005636975.1 | 7 - 8 | 1014 | 151 | |
| XM_005636919.3 | XP_005636976.1 | 9 - 10 | 1154 | 151 | ||
| XM_005636920.2 | XP_005636977.1 | 8 - 9 | 1094 | 151 | ||
Set Membership: |
|