Search
Search
| Gene Symbol: STRADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422823.1 | XP_022278531.1 | 799 | 129 | ||
| XM_022422824.1 | XP_022278532.1 | 903 | 129 | |||
| XM_022422825.1 | XP_022278533.1 | 775 | 129 | |||
| XM_022422826.1 | XP_022278534.1 | 879 | 129 | |||
| XM_022422827.1 | XP_022278535.1 | 606 | 129 | |||
| XM_022422828.1 | XP_022278536.1 | 582 | 129 | |||
| XM_022422829.1 | XP_022278537.1 | 621 | 129 | |||
| XM_855943.5 | XP_861036.2 | 794 | 129 | |||
Set Membership: |
|