Search
Search
| Gene Symbol: CHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113547.2 | XP_013969022.1 | 4 - 5 | 458 | 126 | |
| XM_014113548.2 | XP_013969023.1 | 4 - 5 | 458 | 126 | ||
| XM_014113549.2 | XP_013969024.1 | 4 - 5 | 455 | 126 | ||
| XM_014113550.2 | XP_013969025.1 | 4 - 5 | 538 | 126 | ||
| XM_014113551.2 | XP_013969026.1 | 4 - 5 | 458 | 126 | ||
| XM_014113552.2 | XP_013969027.1 | 4 - 5 | 458 | 126 | ||
| XM_014113553.2 | XP_013969028.1 | 4 - 5 | 458 | 126 | ||
| XM_014113554.2 | XP_013969029.1 | 4 - 5 | 458 | 126 | ||
| XM_014113555.2 | XP_013969030.1 | 4 - 5 | 458 | 126 | ||
| XM_014113556.2 | XP_013969031.1 | 4 - 5 | 458 | 126 | ||
| XM_014113557.2 | XP_013969032.1 | 4 - 5 | 458 | 126 | ||
| XM_014113558.2 | XP_013969033.1 | 4 - 5 | 458 | 126 | ||
| XM_014113559.2 | XP_013969034.1 | 4 - 5 | 458 | 126 | ||
| XM_014113560.2 | XP_013969035.1 | 4 - 5 | 458 | 126 | ||
| XM_014113561.2 | XP_013969036.1 | 4 - 5 | 458 | 126 | ||
| XM_014113562.1 | XP_013969037.1 | 4 - 5 | 458 | 126 | ||
| XM_014113563.2 | XP_013969038.1 | 4 - 5 | 458 | 126 | ||
| XM_014113564.2 | XP_013969039.1 | 4 - 5 | 458 | 126 | ||
Set Membership: |
|