Search
Search
| Gene Symbol: DNM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639728.4 | XP_003639776.1 | 16 - 17 | 2021 | 99 | |
| XM_003639729.4 | XP_003639777.1 | 17 - 18 | 2033 | 99 | ||
| XM_003639730.4 | XP_003639778.1 | 16 - 17 | 2021 | 99 | ||
| XM_005632882.3 | XP_005632939.1 | 18 - 19 | 2063 | 99 | ||
| XM_005632883.3 | XP_005632940.1 | 18 - 19 | 2063 | 99 | ||
| XM_005632884.3 | XP_005632941.1 | 18 - 19 | 2063 | 99 | ||
| XM_005632885.3 | XP_005632942.1 | 17 - 18 | 2051 | 99 | ||
| XM_005632886.3 | XP_005632943.1 | 18 - 19 | 2063 | 99 | ||
| XM_005632887.3 | XP_005632944.1 | 17 - 18 | 2033 | 99 | ||
| XM_005632888.3 | XP_005632945.1 | 17 - 18 | 2033 | 99 | ||
| XM_005632889.3 | XP_005632946.1 | 17 - 18 | 2033 | 99 | ||
| XM_022407050.1 | XP_022262758.1 | 17 - 18 | 2033 | 99 | ||
| XM_022407051.1 | XP_022262759.1 | 17 - 18 | 1744 | 99 | ||
| XM_022407052.1 | XP_022262760.1 | 15 - 16 | 1574 | 99 | ||
| XM_862888.6 | XP_867981.3 | 17 - 18 | 2033 | 99 | ||
Set Membership: |
|