Search
Search
| Gene Symbol: ADAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638326.3 | XP_005638383.1 | 8 - 9 | 997 | 158 | |
| XM_005638327.3 | XP_005638384.1 | 8 - 9 | 971 | 158 | ||
| XM_005638328.3 | XP_005638385.1 | 8 - 9 | 961 | 158 | ||
| XM_005638329.3 | XP_005638386.1 | 8 - 9 | 936 | 158 | ||
| XM_005638330.3 | XP_005638387.1 | 8 - 9 | 1002 | 158 | ||
Set Membership: |
|