Search
Search
| Gene Symbol: GTF2H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639426.4 | XP_003639474.1 | 5 - 6 | 649 | 118 | |
| XM_005627126.3 | XP_005627183.1 | 5 - 6 | 607 | 118 | ||
| XM_022425825.1 | XP_022281533.1 | 5 - 6 | 650 | 118 | ||
Set Membership: |
|