Search
Search
| Gene Symbol: MSI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636302.3 | XP_005636359.1 | 4 - 5 | 342 | 142 | |
| XM_005636303.3 | XP_005636360.1 | 4 - 5 | 343 | 142 | ||
| XM_014108096.2 | XP_013963571.1 | 4 - 5 | 342 | 142 | ||
| XM_022410439.1 | XP_022266147.1 | 4 - 5 | 342 | 142 | ||
| XM_849159.5 | XP_854252.2 | 4 - 5 | 342 | 142 | ||
Set Membership: |
|