Search
Search
| Gene Symbol: GNE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003431575.4 | XP_003431623.1 | 5 - 6 | 1147 | 87 | |
| XM_005626809.3 | XP_005626866.1 | 5 - 6 | 1179 | 87 | ||
| XM_022425601.1 | XP_022281309.1 | 5 - 6 | 1043 | 87 | ||
Set Membership: |
|