Search
Search
| Gene Symbol: IKZF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432287.3 | XP_003432335.2 | 590 | 147 | ||
| XM_003432289.4 | XP_003432337.1 | 971 | 147 | |||
| XM_003432290.3 | XP_003432338.2 | 592 | 147 | |||
| XM_005630898.2 | XP_005630955.1 | 847 | 147 | |||
| XM_005630899.2 | XP_005630956.1 | 850 | 147 | |||
| XM_005630900.2 | XP_005630957.1 | 849 | 147 | |||
| XM_005630901.2 | XP_005630958.1 | 657 | 147 | |||
Set Membership: |
|