Search
Search
| Gene Symbol: PTPN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626910.3 | XP_005626967.1 | 20 - 21 | 2319 | 75 | |
| XM_005626912.3 | XP_005626969.1 | 12 - 13 | 1456 | 75 | ||
| XM_022425695.1 | XP_022281403.1 | 18 - 19 | 1923 | 75 | ||
| XM_022425696.1 | XP_022281404.1 | 17 - 18 | 1755 | 75 | ||
| XM_848423.5 | XP_853516.1 | 19 - 20 | 2180 | 75 | ||
Set Membership: |
|