Search
Search
| Gene Symbol: PTPRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622278.2 | XP_005622335.1 | 3502 | 177 | ||
| XM_005622279.2 | XP_005622336.1 | 3310 | 177 | |||
| XM_005622280.2 | XP_005622337.1 | 3178 | 177 | |||
| XM_005622281.2 | XP_005622338.1 | 3037 | 177 | |||
| XM_005622282.2 | XP_005622339.1 | 2971 | 177 | |||
Set Membership: |
|