Search
Search
| Gene Symbol: SOAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622465.3 | XP_005622522.1 | 14 - 15 | 1479 | 196 | |
| XM_014115153.2 | XP_013970628.1 | 13 - 14 | 1323 | 196 | ||
| XM_547445.6 | XP_547445.2 | 13 - 14 | 1417 | 196 | ||
Set Membership: |
|