Search
Search
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005629550.3 | XP_005629607.1 | 1559 | 163 | ||
| XM_005629551.3 | XP_005629608.1 | 1727 | 163 | |||
| GenBank mRNA | AY545218.2 | - | 1814 | 163 | ||
| AY545219.2 | - | 1894 | 163 | |||
Set Membership: |
|