Search
Search
| Gene Symbol: DYRK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014120454.2 | XP_013975929.2 | 2 - 3 | 1469 | 114 | |
| XM_014120462.2 | XP_013975937.2 | 2 - 3 | 1468 | 114 | ||
| XM_022425706.1 | XP_022281414.1 | 2 - 3 | 1471 | 114 | ||
| XM_541620.5 | XP_541620.2 | 3 - 4 | 436 | 114 | ||
Set Membership: |
|