Search
Search
| Gene Symbol: TPBG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627573.3 | XP_005627630.1 | 2 | 1025 | 152 | |
| XM_005627574.2 | XP_005627631.1 | 2 | 971 | 152 | ||
| XM_022426288.1 | XP_022281996.1 | 2 | 1008 | 152 | ||
Set Membership: |
|