Search
Search
| Gene Symbol: TCERG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617351.3 | XP_005617408.1 | 14 - 15 | 2095 | 110 | |
| XM_005617352.3 | XP_005617409.1 | 13 - 14 | 2032 | 110 | ||
| XM_022409152.1 | XP_022264860.1 | 14 - 15 | 2097 | 110 | ||
| XM_535230.6 | XP_535230.2 | 13 - 14 | 2044 | 110 | ||
| XM_853836.5 | XP_858929.1 | 12 - 13 | 1981 | 110 | ||
Set Membership: |
|