Search
Search
| Gene Symbol: TRIOBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014117368.2 | XP_013972843.1 | 13 - 14 | 5268 | 92 | |
| XM_022424463.1 | XP_022280171.1 | 12 - 13 | 5129 | 92 | ||
| XM_022424464.1 | XP_022280172.1 | 11 - 12 | 5051 | 92 | ||
| XM_022424465.1 | XP_022280173.1 | 13 - 14 | 5272 | 92 | ||
Set Membership: |
|