Search
Search
| Gene Symbol: CNTN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005633312.3 | XP_005633369.1 | 19 - 20 | 2813 | 114 | |
| XM_014122723.2 | XP_013978198.1 | 20 - 21 | 2884 | 114 | ||
| XM_014122727.2 | XP_013978202.1 | 20 - 21 | 2884 | 114 | ||
Set Membership: |
|