Search
Search

ADAMTSL2| Gene Symbol: ADAMTSL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639308.4 | XP_003639356.1 | 6 - 7 | 800 | 109 | |
| XM_005625150.3 | XP_005625207.1 | 6 - 7 | 801 | 109 | ||
| XM_014116955.2 | XP_013972430.1 | 6 - 7 | 959 | 109 | ||
| GenBank mRNA | GU220770.1 | - | 5 - 6 | 554 | 109 | |
Set Membership: |
|