Search
Search
| Gene Symbol: PIEZO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620574.2 | XP_005620631.1 | 2727 | 66 | ||
| XM_022419088.1 | XP_022274796.1 | 2538 | 66 | |||
| XM_022419089.1 | XP_022274797.1 | 2540 | 66 | |||
| XM_022419090.1 | XP_022274798.1 | 2538 | 66 | |||
Set Membership: |
|