Search
Search
| Gene Symbol: PHF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641388.3 | XP_005641445.1 | 16 - 17 | 2803 | 142 | |
| XM_005641389.3 | XP_005641446.1 | 15 - 16 | 2500 | 142 | ||
| XM_005641390.3 | XP_005641447.1 | 15 - 16 | 2499 | 142 | ||
| XM_014111751.2 | XP_013967226.1 | 16 - 17 | 2804 | 142 | ||
| XM_014111752.2 | XP_013967227.1 | 16 - 17 | 2803 | 142 | ||
| XM_022415601.1 | XP_022271309.1 | 15 - 16 | 2612 | 142 | ||
Set Membership: |
|