Search
Search

SLC22A23| Gene Symbol: SLC22A23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639984.3 | XP_005640041.1 | 10 | 1155 | 132 | |
| XM_005639985.3 | XP_005640042.1 | 9 | 1131 | 132 | ||
Set Membership: |
|