Search
Search

TBL1XR1| Gene Symbol: TBL1XR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005639941.3 | XP_005639998.1 | 8 - 9 | 976 | 126 | |
| XM_005639944.3 | XP_005640001.1 | 8 - 9 | 1019 | 126 | ||
| XM_022414178.1 | XP_022269886.1 | 8 - 9 | 1023 | 126 | ||
| XM_022414179.1 | XP_022269887.1 | 8 - 9 | 1105 | 126 | ||
| XM_022414180.1 | XP_022269888.1 | 7 - 8 | 920 | 126 | ||
| XM_022414181.1 | XP_022269889.1 | 7 - 8 | 873 | 126 | ||
Set Membership: |
|