Search
Search

ADAMTS12| Gene Symbol: ADAMTS12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022417949.1 | XP_022273657.1 | 10 - 11 | 1833 | 83 | |
| XM_022417950.1 | XP_022273658.1 | 9 - 10 | 1683 | 83 | ||
| XM_022417951.1 | XP_022273659.1 | 10 - 11 | 1834 | 83 | ||
Set Membership: |
|