Search
Search
| Gene Symbol: HMCN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622446.2 | XP_005622503.1 | 105 | 16203 | 118 | |
| XM_005622447.2 | XP_005622504.1 | 96 | 14837 | 118 | ||
| XM_022420976.1 | XP_022276684.1 | 105 | 16272 | 118 | ||
| XM_547438.5 | XP_547438.2 | 106 | 16554 | 118 | ||
Set Membership: |
|