Search
Search
| Gene Symbol: NCALD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627835.3 | XP_005627892.1 | 459 | 141 | ||
| XM_005627836.3 | XP_005627893.1 | 526 | 141 | |||
| XM_022426593.1 | XP_022282301.1 | 316 | 141 | |||
| XM_022426594.1 | XP_022282302.1 | 343 | 141 | |||
| XM_022426595.1 | XP_022282303.1 | 290 | 141 | |||
| XM_022426596.1 | XP_022282304.1 | 404 | 141 | |||
| XM_022426597.1 | XP_022282305.1 | 421 | 141 | |||
Set Membership: |
|