Search
Search
| Gene Symbol: MCM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005634804.3 | XP_005634861.1 | 7 - 8 | 1249 | 151 | |
| XM_014106912.2 | XP_013962387.1 | 7 - 8 | 1171 | 151 | ||
| XM_022409212.1 | XP_022264920.1 | 7 - 8 | 1381 | 151 | ||
| XM_022409213.1 | XP_022264921.1 | 8 - 9 | 906 | 151 | ||
| XM_022409214.1 | XP_022264922.1 | 8 - 9 | 904 | 151 | ||
Set Membership: |
|