Search
Search
| Gene Symbol: PSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022425439.1 | XP_022281147.1 | 5 - 6 | 574 | 102 | |
| XM_022425440.1 | XP_022281148.1 | 5 - 6 | 574 | 102 | ||
| XM_022425441.1 | XP_022281149.1 | 5 - 6 | 574 | 102 | ||
| XM_859602.5 | XP_864695.1 | 5 - 6 | 574 | 102 | ||
Set Membership: |
|