Search
Search
| Gene Symbol: KCNH7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110763.2 | XP_013966238.1 | 7 - 8 | 1727 | 168 | |
| XM_022414824.1 | XP_022270532.1 | 7 - 8 | 1727 | 168 | ||
| XM_022414825.1 | XP_022270533.1 | 6 - 7 | 1706 | 168 | ||
Set Membership: |
|