Search
Search
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002998.1 | NP_001002998.1 | 3 | 749 | 136 | |
| XM_014121722.2 | XP_013977197.1 | 2 | 776 | 136 | ||
| XM_014121724.2 | XP_013977199.1 | 9 | 1619 | 136 | ||
| GenBank mRNA | AB104899.3 | - | 3 | 749 | 136 | |
| AY859723.1 | - | 2 | 687 | 136 | ||
Set Membership: |
|