Search
Search
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416776.1 | XP_022272484.1 | 1763 | 115 | ||
| XM_022416777.1 | XP_022272485.1 | 1627 | 115 | |||
| XM_022416779.1 | XP_022272487.1 | 1535 | 115 | |||
| XM_022416780.1 | XP_022272488.1 | 1673 | 115 | |||
| XM_022416781.1 | XP_022272489.1 | 1670 | 115 | |||
| XM_536320.6 | XP_536320.3 | 1760 | 115 | |||
Set Membership: |
|