Search
Search
| Gene Symbol: SLC9C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622515.2 | XP_005622572.1 | 22 - 23 | 2955 | 180 | |
| XM_005622517.2 | XP_005622574.1 | 21 - 22 | 2667 | 180 | ||
| XM_022421034.1 | XP_022276742.1 | 22 - 23 | 2942 | 180 | ||
| XM_022421035.1 | XP_022276743.1 | 22 - 23 | 2967 | 180 | ||
| XM_022421036.1 | XP_022276744.1 | 22 - 23 | 2926 | 180 | ||
| XM_022421037.1 | XP_022276745.1 | 22 - 23 | 2836 | 180 | ||
| XM_022421039.1 | XP_022276747.1 | 21 - 22 | 2850 | 180 | ||
| XM_022421040.1 | XP_022276748.1 | 21 - 22 | 2835 | 180 | ||
| XM_022421041.1 | XP_022276749.1 | 22 - 23 | 2955 | 180 | ||
| XM_022421042.1 | XP_022276750.1 | 21 - 22 | 2766 | 180 | ||
| XM_022421043.1 | XP_022276751.1 | 20 - 21 | 2730 | 180 | ||
| XM_022421044.1 | XP_022276752.1 | 19 - 20 | 2601 | 180 | ||
| XM_022421045.1 | XP_022276753.1 | 18 - 19 | 2481 | 180 | ||
Set Membership: |
|