Search
Search
| Gene Symbol: DHX57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630362.3 | XP_005630419.1 | 20 - 21 | 3773 | 98 | |
| XM_005630363.3 | XP_005630420.1 | 21 - 22 | 3911 | 98 | ||
| XM_014120332.2 | XP_013975807.1 | 19 - 20 | 3526 | 98 | ||
| XM_014120333.2 | XP_013975808.1 | 18 - 19 | 3278 | 98 | ||
| XM_540155.6 | XP_540155.1 | 20 - 21 | 3673 | 98 | ||
Set Membership: |
|