Search
Search

TBC1D22A| Gene Symbol: TBC1D22A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625744.2 | XP_005625801.1 | 1 - 2 | 226 | 58 | |
| XM_022424283.1 | XP_022279991.1 | 1 - 2 | 226 | 58 | ||
| XM_843497.4 | XP_848590.1 | 1 - 2 | 225 | 58 | ||
Set Membership: |
|