Search
Search

TXNDC11| Gene Symbol: TXNDC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621534.3 | XP_005621591.1 | 9 - 10 | 1981 | 154 | |
| XM_005621536.3 | XP_005621593.1 | 7 - 8 | 1656 | 154 | ||
| XM_005621537.3 | XP_005621594.1 | 7 - 8 | 1528 | 154 | ||
| XM_022420058.1 | XP_022275766.1 | 9 - 10 | 1893 | 154 | ||
| XM_022420059.1 | XP_022275767.1 | 7 - 8 | 1665 | 154 | ||
| XM_022420060.1 | XP_022275768.1 | 7 - 8 | 1792 | 154 | ||
| XM_022420061.1 | XP_022275769.1 | 7 - 8 | 1496 | 154 | ||
| XM_022420062.1 | XP_022275770.1 | 9 - 10 | 1833 | 154 | ||
Set Membership: |
|