Search
Search
| Gene Symbol: PTCD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630475.3 | XP_005630532.1 | 19 - 20 | 1609 | 142 | |
| XM_005630476.2 | XP_005630533.2 | 19 - 20 | 1681 | 142 | ||
| XM_005630477.3 | XP_005630534.1 | 17 - 18 | 1407 | 142 | ||
| XM_014120363.2 | XP_013975838.1 | 16 - 17 | 1400 | 142 | ||
| XM_532975.6 | XP_532975.3 | 19 - 20 | 1599 | 142 | ||
Set Membership: |
|