Search
Search

SPATA20| Gene Symbol: SPATA20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005624644.3 | XP_005624701.1 | 2026 | 125 | ||
| XM_005624645.3 | XP_005624702.1 | 1930 | 125 | |||
| XM_022423102.1 | XP_022278810.1 | 1844 | 125 | |||
Set Membership: |
|