Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HYI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022428067.1 | XP_022283775.1 | 4 - 5 | 637 | 143 | |
XM_539648.6 | XP_539648.4 | 4 - 5 | 568 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Canis familiaris Probe spans exons |