Search
Search
| Gene Symbol: LOXL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630521.3 | XP_005630578.1 | 13 - 14 | 2203 | 81 | |
| XM_022404781.1 | XP_022260489.1 | 13 - 14 | 2195 | 81 | ||
| XM_022404782.1 | XP_022260490.1 | 6 - 7 | 974 | 81 | ||
| XM_847431.5 | XP_852524.1 | 11 - 12 | 2086 | 81 | ||
Set Membership: |
|