Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Cf02727364_m1
          See other PTPRD GE Assays ›
          Gene Symbol:
          PTPRD
          Gene Name:
          protein tyrosine phosphatase receptor type D
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 29100913 - 31278495 on Build CanFam3.1
          Species:
          Dog
          Species Specific ID (Flybase ID):
          -
          Assay ID Cf02727364_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: PTPRD
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_005626620.3 XP_005626677.1 45 - 46 6531 163
          XM_014117752.2 XP_013973227.1 40 - 41 6414 163
          XM_014117753.2 XP_013973228.1 45 - 46 6522 163
          XM_014117756.2 XP_013973231.1 37 - 38 5218 163
          XM_014117757.2 XP_013973232.1 36 - 37 5206 163
          XM_022425377.1 XP_022281085.1 45 - 46 6534 163
          XM_022425378.1 XP_022281086.1 44 - 45 6479 163
          XM_022425379.1 XP_022281087.1 45 - 46 6531 163
          XM_022425380.1 XP_022281088.1 45 - 46 6531 163
          XM_022425381.1 XP_022281089.1 45 - 46 6532 163
          XM_022425382.1 XP_022281090.1 45 - 46 6529 163
          XM_022425383.1 XP_022281091.1 44 - 45 6525 163
          XM_022425384.1 XP_022281092.1 44 - 45 6522 163
          XM_022425385.1 XP_022281093.1 44 - 45 6519 163
          XM_022425386.1 XP_022281094.1 44 - 45 6516 163
          XM_022425387.1 XP_022281095.1 44 - 45 6513 163
          XM_022425388.1 XP_022281096.1 43 - 44 6507 163
          XM_022425389.1 XP_022281097.1 44 - 45 6507 163
          XM_022425390.1 XP_022281098.1 43 - 44 6504 163
          XM_022425391.1 XP_022281099.1 42 - 43 6495 163
          XM_022425392.1 XP_022281100.1 42 - 43 6492 163
          XM_022425393.1 XP_022281101.1 44 - 45 6492 163
          XM_022425394.1 XP_022281102.1 44 - 45 6489 163
          XM_022425395.1 XP_022281103.1 44 - 45 6486 163
          XM_022425396.1 XP_022281104.1 43 - 44 6477 163
          XM_022425397.1 XP_022281105.1 43 - 44 6474 163
          XM_022425398.1 XP_022281106.1 43 - 44 6470 163
          XM_022425399.1 XP_022281107.1 41 - 42 6462 163
          XM_022425400.1 XP_022281108.1 39 - 40 6404 163
          XM_022425401.1 XP_022281109.1 44 - 45 6228 163
          XM_022425402.1 XP_022281110.1 43 - 44 6216 163
          XM_022425403.1 XP_022281111.1 42 - 43 6204 163
          XM_022425404.1 XP_022281112.1 41 - 42 6186 163
          XM_022425405.1 XP_022281113.1 40 - 41 6158 163
          XM_022425406.1 XP_022281114.1 38 - 39 5266 163
          XM_022425407.1 XP_022281115.1 39 - 40 5251 163
          XM_022425408.1 XP_022281116.1 39 - 40 5248 163
          XM_022425409.1 XP_022281117.1 37 - 38 5248 163
          XM_022425410.1 XP_022281118.1 38 - 39 5239 163
          XM_022425411.1 XP_022281119.1 38 - 39 5239 163
          XM_022425412.1 XP_022281120.1 38 - 39 5236 163
          XM_022425413.1 XP_022281121.1 37 - 38 5221 163
          XM_022425414.1 XP_022281122.1 38 - 39 6101 163

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Recommended Assay Canis familiaris Probe spans exons

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline