Search
Search

PPARGC1B| Gene Symbol: PPARGC1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619306.3 | XP_005619363.1 | 5 - 6 | 1796 | 105 | |
| XM_546308.6 | XP_546308.3 | 5 - 6 | 1796 | 105 | ||
Set Membership: |
|